(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid 95%

Supplier: Ambeed
Warning

A576707-100MG A576707-1G A576707-250MG A576707-5G
77034-982EA 35 USD
77034-982 77034-984 77034-986 77034-988
(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid 95%
(4-Methyl-3-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MDL Number: MFCD09832293
CAS Number: 1000544-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR