1-(4-Nitrophenyl)cyclopropanecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A576300-100MG
A576300-1G
A576300-250MG
A576300-5G
76991-850EA
21.22
USD
76991-850
76991-852
76991-854
76991-856
1-(4-Nitrophenyl)cyclopropanecarboxylic acid 98%
1-(4-Nitrophenyl)cyclopropanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₄ MW: 207.1828 g/mol |
MDL Number:
MFCD07374438 CAS Number: 23348-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...