6-Oxo-1,6-dihydropyridine-3-carbonitrile 98%

Supplier: Ambeed
Warning

A576170-5G A576170-1G
77034-980EA 165.51 USD
77034-980 77594-748
6-Oxo-1,6-dihydropyridine-3-carbonitrile 98%
6-Hydroxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄N₂O
MW: 120.109 g/mol
MDL Number: MFCD07698589
CAS Number: 94805-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR