N-Benzyl-N-methylglycine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzyl(methyl)amino)acetic acid

A575654-1G A575654-250MG
77017-820EA 306 USD
77017-820 77017-822
N-Benzyl-N-methylglycine 95%
N-Benzyl-N-methylglycine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD09711201
CAS Number: 37429-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR