8-Benzyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one 96%

Supplier: Ambeed
Warning

A574738-250MG A574738-1G
76991-844EA 41 USD
76991-844 77886-398
8-Benzyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one 96%
8-Benzyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₂
MDL Number: MFCD01710744
CAS Number: 5053-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR