2,2-Dimethylcyclopentanol 98%

Supplier: Ambeed
Danger

A574523-250MG A574523-100MG A574523-1G
76991-842EA 31 USD
76991-842 77891-145 77891-146
2,2-Dimethylcyclopentanol 98%
2,2-Dimethylcyclopentanol

Precaución: for Research Use Only

Formula: C₇H₁₄O
MDL Number: MFCD11655617
CAS Number: 37617-33-7
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR