1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A574228-100MG A574228-1G
77005-290EA 28.65 USD
77005-290 77005-292
1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid 97%
1H-Pyrrolo[3,2-b]pyridine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD09965906
CAS Number: 872355-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR