2-Bromo-N,N-dimethylacetamide 98%

Supplier: Ambeed
Danger

A573966-10G A573966-1G A573966-25G A573966-5G
77005-282EA 19 USD
77005-282 77005-284 77005-286 77005-288
2-Bromo-N,N-dimethylacetamide 98%
2-Bromo-N,N-dimethylacetamide

Precaución: for Research Use Only

Formula: C₄H₈BrNO
MDL Number: MFCD12026683
CAS Number: 5468-77-9
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR