D-N-Benzylserine methyl ester 96%

Supplier: Ambeed
Warning

A573870-100MG A573870-1G
77005-278EA 26 USD
77005-278 77005-280
D-N-Benzylserine methyl ester 96%
D-N-Benzylserine methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MDL Number: MFCD12198843
CAS Number: 131110-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR