5-Iodo-1H-pyrrolo[2,3-b]pyridine 96%

Supplier: Ambeed
Warning

A573730-1G A573730-250MG
77017-810EA 149.59 USD
77017-810 77017-812
5-Iodo-1H-pyrrolo[2,3-b]pyridine 96%
5-Iodo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MDL Number: MFCD06659679
CAS Number: 898746-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR