4-(2-Methoxyphenoxy)aniline 98%

Supplier: Ambeed
Warning

A573590-100MG A573590-1G A573590-250MG A573590-5G
77034-972EA 16.98 USD
77034-972 77034-974 77034-976 77034-978
4-(2-Methoxyphenoxy)aniline 98%
4-(2-Methoxyphenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
MDL Number: MFCD02663341
CAS Number: 13066-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR