N,N'-Bis(dimethylaminomethylene)hydrazine 98%

Supplier: Ambeed
Warning

A573478-250MG A573478-1G A573478-5G
76991-828EA 25 USD
76991-828 77891-142 77891-143
N,N'-Bis(dimethylaminomethylene)hydrazine 98%
N,N'-Bis(dimethylaminomethylene)hydrazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₄
MDL Number: MFCD00957712
CAS Number: 16114-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR