5-Hydroxyisobenzofuran-1(3H)-one 95%

Supplier: Ambeed
Warning

A570979-100MG A570979-1G A570979-250MG A570979-25G A570979-5G
76991-808EA 15 USD
76991-808 76991-810 76991-812 76991-814 76991-816
5-Hydroxyisobenzofuran-1(3H)-one 95%
5-Hydroxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₆O₃
MDL Number: MFCD03425897
CAS Number: 55104-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR