6-Iodo-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A570103-1G A570103-250MG
77005-270EA 201.58 USD
77005-270 77005-272
6-Iodo-1H-pyrrolo[3,2-b]pyridine 95%
6-Iodo-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MDL Number: MFCD09859119
CAS Number: 1015609-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR