trans-2-Octenal 98%

Supplier: Ambeed
Warning

Synonyms: (E)-2-Octenal

A569791-100G A569791-25G A569791-500G A569791-5G
77005-262EA 97.61 USD
77005-262 77005-264 77005-266 77005-268
trans-2-Octenal 98%
trans-2-Octenal

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.2 g/mol
MDL Number: MFCD00007011
CAS Number: 2548-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR