3-Hydroxy-4-methylbenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Borono-2-methylphenol, (3-Hydroxy-4-methylphenyl)boronic acid, 3-Hydroxy-4-methylphenylboronic acid, 5-Borono-o-cresol, 4-Borono-2-hydroxytoluene

A569737-100MG A569737-1G A569737-250MG
77034-954EA 33.95 USD
77034-954 77034-956 77034-958
3-Hydroxy-4-methylbenzeneboronic acid 98%
3-Hydroxy-4-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.956 g/mol
MDL Number: MFCD01074652
CAS Number: 216019-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR