Dimethylphosphine oxide 95%

Supplier: Ambeed
Danger

A569270-10G A569270-1G A569270-25G A569270-5G
77005-254EA 51.99 USD
77005-254 77005-256 77005-258 77005-260
Dimethylphosphine oxide 95%
Dimethylphosphine oxide

Precaución: for Research Use Only

Formula: C₂H₇OP
MW: 78.05 g/mol
MDL Number: MFCD02824571
CAS Number: 7211-39-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR