4-Bromo-2-(tert-butyl)pyridine 97%

Supplier: Ambeed
Warning

A567812-100MG A567812-1G A567812-250MG
76991-798EA 124.13 USD
76991-798 76991-800 76991-802
4-Bromo-2-(tert-butyl)pyridine 97%
4-Bromo-2-(tert-butyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MW: 214.10 g/mol
MDL Number: MFCD11223211
CAS Number: 1086381-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR