(6-Phenylnaphthalen-2-yl)boronic acid 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Phenylnaphthalene-2-boronic acid
A567435-100MG
A567435-1G
A567435-250MG
77017-784EA
8
USD
77017-784
77017-786
77017-788
(6-Phenylnaphthalen-2-yl)boronic acid 95+%
(6-Phenylnaphthalen-2-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₃BO₂ |
MDL Number:
MFCD26406213 CAS Number: 876442-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...