[Methyl(phenyl)amino](oxo)acetic acid 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Methyl(phenyl)amino)-2-oxoacetic acid

A567282-100MG A567282-1G A567282-250MG
77034-942EA 40.32 USD
77034-942 77034-944 77034-946
[Methyl(phenyl)amino](oxo)acetic acid 95%
[Methyl(phenyl)amino](oxo)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD09864583
CAS Number: 13120-33-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR