2-(Boc-amino)-2-(3,4-dichlorophenyl)acetic Acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A567217-1G A567217-5G
77017-782EA 51 USD
77017-782 77684-422
2-(Boc-amino)-2-(3,4-dichlorophenyl)acetic Acid 95%, Ambeed.Inc
2-(Boc-amino)-2-(3,4-dichlorophenyl)acetic Acid

Precaución: For research use only.

Formula: C₁₃H₁₅Cl₂NO₄
MW: 320.17 g/mol
MDL Number: MFCD03425232
CAS Number: 142121-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR