alpha-(3-Bromophenyl)benzylamine 97%

Supplier: Ambeed
Warning

A567135-5G A567135-1G A567135-250MG
77017-780EA 189 USD
77017-780 77891-131 77891-132
alpha-(3-Bromophenyl)benzylamine 97%
alpha-(3-Bromophenyl)benzylamine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN
MW: 262.14 g/mol
MDL Number: MFCD09738398
CAS Number: 55095-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR