Methyl 4-formyl-3-methoxybenzoate 97%

Supplier: Ambeed
Warning

A567086-1G A567086-250MG A567086-10G
77017-776EA 40 USD
77017-776 77017-778 77891-130
Methyl 4-formyl-3-methoxybenzoate 97%
Methyl 4-formyl-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
MDL Number: MFCD17169951
CAS Number: 74733-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR