2-Bromo-4,5-dimethylphenol 95%

Supplier: Ambeed
Warning

A567023-250MG A567023-5G
76991-794EA 50 USD
76991-794 76991-796
2-Bromo-4,5-dimethylphenol 95%
2-Bromo-4,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD09260834
CAS Number: 22802-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR