2,2'-Methylenebis(4-aminophenol) dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A566831-100G
A566831-25G
A566831-500G
A566831-5G
77005-238EA
36.08
USD
77005-238
77005-240
77005-242
77005-244
2,2'-Methylenebis(4-aminophenol) dihydrochloride 97%
2,2'-Methylenebis(4-aminophenol) dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₆Cl₂N₂O₂ MW: 303.18 g/mol |
MDL Number:
MFCD07368625 CAS Number: 27311-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...