2-Chloro-4-iodonicotinonitrile 98%

Supplier: Ambeed
Warning

A566030-1G A566030-250MG A566030-25G A566030-5G
77005-226EA 21.22 USD
77005-226 77005-228 77005-230 77005-232
2-Chloro-4-iodonicotinonitrile 98%
2-Chloro-4-iodonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₂ClIN₂
MDL Number: MFCD12401632
CAS Number: 1171919-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR