5-Chloro-2-iodo-4-methylpyrimidine 95%

Supplier: Ambeed
Warning

A565503-100MG A565503-1G A565503-250MG
77017-766EA 46.68 USD
77017-766 77017-768 77017-770
5-Chloro-2-iodo-4-methylpyrimidine 95%
5-Chloro-2-iodo-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MW: 254.45 g/mol
MDL Number: MFCD13193657
CAS Number: 1260810-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR