tert-Butyl 3-(trifluoromethyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A564989-100MG A564989-250MG A564989-50MG A564989-1G
76991-782EA 84 USD
76991-782 76991-784 76991-786 77569-086
tert-Butyl 3-(trifluoromethyl)piperazine-1-carboxylate 95%
tert-Butyl 3-(trifluoromethyl)piperazine-1-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₇F₃N₂O₂
MW: 254.24 g/mol
MDL Number: MFCD08686626
CAS Number: 886779-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR