6-Chloro-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: Ambeed
Danger

Synonyms: 6-Chloro-4-azaindole

A564512-100MG A564512-1G A564512-250MG A564512-5G
77034-930EA 37.14 USD
77034-930 77034-932 77034-934 77034-936
6-Chloro-1H-pyrrolo[3,2-b]pyridine 97%
6-Chloro-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD10574982
CAS Number: 1021339-19-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR