2-Bromo-4-(trifluoromethyl)benzyl chloride 98+%

Supplier: Ambeed
Danger

A564191-1G A564191-250MG A564191-5G
76991-774EA 29 USD
76991-774 77886-380 77886-381
2-Bromo-4-(trifluoromethyl)benzyl chloride 98+%
2-Bromo-4-(trifluoromethyl)benzyl chloride

Precaución: for Research Use Only

Formula: C₈H₅BrClF₃
MDL Number: MFCD03094249
CAS Number: 480438-96-8
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR