(S)-1-Boc-2-benzylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-benzylpiperazine-1-carboxylate

A564015-1G A564015-250MG
77017-754EA 102.91 USD
77017-754 77017-756
(S)-1-Boc-2-benzylpiperazine 97%
(S)-1-Boc-2-benzylpiperazine

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂O₂
MW: 276.38 g/mol
MDL Number: MFCD03787923
CAS Number: 169447-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR