tert-Butyl 1-oxo-2,9-diazaspiro[5.5]undecane-9-carboxylate 98%

Supplier: Ambeed
Warning

A563525-100MG A563525-1G A563525-250MG A563525-5G
76991-766EA 9 USD
76991-766 76991-768 76991-770 76991-772
tert-Butyl 1-oxo-2,9-diazaspiro[5.5]undecane-9-carboxylate 98%
tert-Butyl 1-oxo-2,9-diazaspiro[5.5]undecane-9-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₄N₂O₃
MDL Number: MFCD13191664
CAS Number: 1198284-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR