4'-n-Propoxybiphenyl-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4'-Propoxy-[1,1'-biphenyl]-4-yl)boronic acid

A563198-1G A563198-250MG A563198-5G
76991-760EA 126.25 USD
76991-760 76991-762 76991-764
4'-n-Propoxybiphenyl-4-boronic acid 98%
4'-n-Propoxybiphenyl-4-boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₇BO₃
MW: 256.108 g/mol
MDL Number: MFCD06798061
CAS Number: 849062-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR