6-(Hydroxymethyl)indole 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (1H-Indol-6-yl)methanol

A562554-100MG A562554-5G A562554-250MG
77017-748EA 22.28 USD
77017-748 77017-750 77684-414
6-(Hydroxymethyl)indole 98%, Ambeed.Inc
6-(Hydroxymethyl)indole

Precaución: For research use only.

Formula: C₉H₉NO
MW: 147.1739 g/mol
MDL Number: MFCD02179595
CAS Number: 1075-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR