Methyl 4-methoxy-3-methylbenzoate 98%

Supplier: Ambeed
Warning

A562449-1G A562449-250MG A562449-5G
77005-198EA 10 USD
77005-198 77005-200 77005-202
Methyl 4-methoxy-3-methylbenzoate 98%
Methyl 4-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD06203809
CAS Number: 70347-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR