Losartan potassium 98%

Supplier: Ambeed
Danger

A561984-10G A561984-1G A561984-5G A561984-25G
77005-186EA 10.61 USD
77005-186 77005-188 77005-190 77673-502
Losartan potassium 98%
Losartan potassium

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClN₆O
MDL Number: MFCD02092704
CAS Number: 124750-99-8
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR