[1,2,4]Triazolo[1,5-a]pyridin-2-amine 97%

Supplier: Ambeed
Warning

A561444-100MG A561444-1G A561444-250MG
77034-912EA 19 USD
77034-912 77034-914 77669-490
[1,2,4]Triazolo[1,5-a]pyridin-2-amine 97%
[1,2,4]Triazolo[1,5-a]pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD11846603
CAS Number: 874-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR