N-(2-Methoxybenzyl)-2-propanamine 98%

Supplier: Ambeed
Warning

A560730-100MG A560730-250MG A560730-5G A560730-1G
77005-174EA 11 USD
77005-174 77005-176 77005-178 77882-347
N-(2-Methoxybenzyl)-2-propanamine 98%
N-(2-Methoxybenzyl)-2-propanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MDL Number: MFCD08691655
CAS Number: 756474-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR