2,6-Difluoro-4-hydroxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2,6-Difluoro-4-hydroxyphenyl)boronic acid

A559997-1G A559997-250MG A559997-5G
77005-168EA 92.3 USD
77005-168 77005-170 77005-172
2,6-Difluoro-4-hydroxyphenylboronic acid 97%
2,6-Difluoro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD09878344
CAS Number: 957065-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR