4-Chloro-2-fluoro-3-methoxybenzoic acid 95%

Supplier: Ambeed
Warning

A559920-250MG A559920-1G
77017-736EA 27.59 USD
77017-736 77684-411
4-Chloro-2-fluoro-3-methoxybenzoic acid 95%
4-Chloro-2-fluoro-3-methoxybenzoic acid

Precaución: For research use only.

Formula: C₈H₆ClFO₃
MW: 204.59 g/mol
MDL Number: MFCD19687204
CAS Number: 1169870-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR