2-Butoxy-5-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

A558883-100MG A558883-1G A558883-250MG A558883-5G
77034-900EA 14 USD
77034-900 77034-902 77034-904 77034-906
2-Butoxy-5-methylphenylboronic acid 98%
2-Butoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
MDL Number: MFCD04974068
CAS Number: 480438-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR