(S)-3-(Boc-amino)-3-(3-methylphenyl)propionic acid 98+%

Supplier: Ambeed
Warning

A558525-100MG A558525-1G A558525-250MG
77034-894EA 8 USD
77034-894 77034-896 77034-898
(S)-3-(Boc-amino)-3-(3-methylphenyl)propionic acid 98+%
(S)-3-(Boc-amino)-3-(3-methylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD03427896
CAS Number: 499995-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR