2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxamide

A558043-250MG A558043-1G A558043-5G
77034-892EA 14.86 USD
77034-892 77673-488 77673-490
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide 95%
2-Amino-6-methyl-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid amide

Precaución: for Research Use Only

Formula: C₉H₁₃N₃OS
MW: 211.29 g/mol
MDL Number: MFCD00687506
CAS Number: 24248-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR