N-Carbobenzoxyglycine 4-nitrophenyl ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-Glycine-p-Nitrophenyl Ester, 4-Nitrophenyl ((benzyloxy)carbonyl)glycinate, Z-Gly-ONp, N-Cbz-glycine 4-Nitrophenyl Ester

A557935-100G A557935-25G A557935-5G
77005-156EA 349.04 USD
77005-156 77005-158 77005-160
N-Carbobenzoxyglycine 4-nitrophenyl ester 97%
N-Carbobenzoxyglycine 4-nitrophenyl ester

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂O₆
MW: 330.30 g/mol
MDL Number: MFCD00024663
CAS Number: 1738-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR