5-Bromo-2,3-dimethoxypyridine 97%

Supplier: Ambeed
Warning

A557906-100MG A557906-1G A557906-250MG A557906-25G A557906-5G
77005-146EA 32.89 USD
77005-146 77005-148 77005-150 77005-152 77005-154
5-Bromo-2,3-dimethoxypyridine 97%
5-Bromo-2,3-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MDL Number: MFCD11052853
CAS Number: 52605-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR