Fmoc-3-(1-naphthyl)-D-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-1-Nal-OH, N-Fmoc-3-(1-naphthyl)-D-alanine

A557736-100G A557736-1G A557736-250MG A557736-25G
77005-138EA 1177.6 USD
77005-138 77005-140 77005-142 77005-144
Fmoc-3-(1-naphthyl)-D-alanine 98%
Fmoc-3-(1-naphthyl)-D-alanine

Precaución: for Research Use Only

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
MDL Number: MFCD00191200
CAS Number: 138774-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR