3-Amino-6-chloro-1H-indazole 97%

Supplier: Ambeed
Warning

A557063-1G A557063-250MG A557063-5G A557063-10G
77034-882EA 60.48 USD
77034-882 77034-884 77034-886 77669-472
3-Amino-6-chloro-1H-indazole 97%
3-Amino-6-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.6000 g/mol
MDL Number: MFCD00047207
CAS Number: 16889-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR