2-(Methylsulfonyl)thiophene 98%

Supplier: Ambeed
Warning

A556414-250MG A556414-5G
77017-716EA 11 USD
77017-716 77017-718
2-(Methylsulfonyl)thiophene 98%
2-(Methylsulfonyl)thiophene

Precaución: for Research Use Only

Formula: C₅H₆O₂S₂
MDL Number: MFCD00052826
CAS Number: 38695-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR