(3-((Pyridin-4-ylthio)methyl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A555844-100MG A555844-1G A555844-250MG
76991-724EA 45.62 USD
76991-724 76991-726 76991-728
(3-((Pyridin-4-ylthio)methyl)phenyl)methanol 95%
(3-((Pyridin-4-ylthio)methyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₃NOS
MW: 231.31 g/mol
MDL Number: MFCD20484656
CAS Number: 811801-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR