Ethyl-3-chloro-4-iodobenzoate 98+%

Supplier: Ambeed
Warning

A555653-100MG A555653-1G A555653-250MG A555653-5G
76991-716EA 5 USD
76991-716 76991-718 76991-720 76991-722
Ethyl-3-chloro-4-iodobenzoate 98+%
Ethyl 3-chloro-4-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClIO₂
MDL Number: MFCD06637384
CAS Number: 874831-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR